Thianthrene
Catalog # PROONIST-1656
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:30 g
- Seq:3332
Specifications
About this item
:
:216.33 g/mol
EINECS:202-197-0
CAS Number:92-85-3
Formula:C₁₂H₈S₂
MDL Number:MFCD00005065