Dichlorvos-[dimethyl-D6] (2,2-Dichlorovinyl dimethyl phosphate-[dimethyl-D6], DDVP-[dimethyl-D6]) 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA12530100CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1,1 mL
- CAS No.:203645-53-8
- Description:Dichlorvos-[dimethyl-D6] (2,2-Dichlorovinyl dimethyl phosphate-[dimethyl-D6], DDVP-[dimethyl-D6]) 100 µg/ml in cyclohexane
- Formula:C₄HD₆Cl₂O₄P
- Description:Dichlorvos-[dimethyl-D6] (2,2-Dichlorovinyl dimethyl phosphate-[dimethyl-D6], DDVP-[dimethyl-D6])