Sarcosine methyl ester hydrochloride
Catalog # BACE4001024.0025
Supplier: BACHEM BIOCHEMICA
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:139.58 g/mol
EINECS:236-853-2
CAS Number:13515-93-0
Formula:C₄H₉NO₂·HCl
MDL Number:MFCD00038876