Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6,7,8-Tetrahydro-2-naphthol
5,6,7,8-Tetrahydro-2-naphthol, Sigma-Aldrich®
5,6,7,8-Tetrahydro-2-naphthol, Sigma-Aldrich®
5,6,7,8-Tetrahydro-2-naphthol, Sigma-Aldrich®
SIAL180459-10G
:
Merck
:
5,6,7,8-Tetrahydro-2-naphthol, Sigma-Aldrich®
SIAL180459-10G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
10 g
Seq:
3236
:
:
148.2 g/mol
:
M-Clarity™ quality level = MQ100
:
214-410-4
:
1125-78-6
:
C₁₀H₁₂O
:
MFCD00001738