Order Entry
United Kingdom
ContactUsLinkComponent
Dinoseb (2-sec-butyl-4,6-dinitrophenol)
Dinoseb (2-sec-butyl-4,6-dinitrophenol)
Catalog # EHERC12810000
Supplier:  EHRENSTORFER
CAS Number:  
Dinoseb (2-sec-butyl-4,6-dinitrophenol)
Catalog # EHERC12810000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020
  • Environmentally Preferable:

Specifications

About this item

 :
 :240.22 g/mol
EINECS:201-861-7
UN:2811
CAS Number:88-85-7
Formula:C₁₀H₁₂N₂O₅
ADR:6.1 II