Order Entry
United Kingdom
ContactUsLinkComponent
Yttrium (III) nitrate hexahydrate ≥99.9% (Y basis)
Oxidizing
Irritant
Yttrium (III) nitrate hexahydrate ≥99.9% (Y basis)
Catalog # ABCRAB122290.1000
Supplier:  abcr
CAS Number:  
Oxidizing
Irritant
Yttrium (III) nitrate hexahydrate ≥99.9% (Y basis)
Catalog # ABCRAB122290.1000
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

About this item

 :
 :383.01 g/mol
Melting Pt:51.8 °C
Density:2.682 g/cm³ (20 °C)
EINECS:233-802-6
UN:1477
ADR:5.1,II
 :GHS03
 :GHS07
CAS Number:13494-98-9
Formula:N₃O₉Y·6H₂O
MDL Number:MFCD00149940