Specifications
- Pk:1 KIT
- Description:L-Lactic acid
- Environmentally Preferable:
- Tests:33
Specifications
About this item
Enzymatic methods allow the rapid, reliable determination of several food constituents. The reagents of the kit have a high stability. For this reason no cooling is necessary during transport. Many test kits contain enzymatic reagents in appropriate quantities for a number of specific determinations eg acetaldehyde kit contains three sets of reagents to carry out 3×11 determinations.
- Reagents are ready to use
- Average expiry of test kit is six months
- Many kits contain sets of reagents to ensure flexible use
More enzymatic test kits on request.
Delivery: Each kit is supplied with a pack insert that provides instructions including wavelength data and also application examples.