Order Entry
United Kingdom
ContactUsLinkComponent
VWR® Smoothie™, Pipette Filler
VWR® Smoothie™, Pipette Filler
Catalog # 612-4546
Supplier:  VWR Collection
undefined
VWR® Smoothie™, Pipette Filler
Catalog # 612-4546
Supplier:  VWR Collection
Specifications
  • Description:
    Pipette filler Smoothie™
  • Pk:
    1
Specifications
About this item
Pipette filler for glass and plastic pipettes from 1 to 25 ml.
  • Comfortable and simple to use
  • Smooth, controllable thumb action
  • Silicone adapter for leakproof positioning of each pipette
The Smoothie™ pipette filler is a simple manual pipetting aid for convenient single handed operation. The unique range control allows selection from a choice of two dispense resolutions. One range setting delivers fast filling with optimal accuracy when used with 10 to 25 ml pipettes, the other setting is for use with pipettes from 1 to 10 ml where accuracy of fill can sometimes be difficult to control.