Conductivity cells
Conductivity probe HI 76309, 4-ring, built-in temperature sensor, ATC, HI 76309, 4-ring, built-in temperature sensor, Stainless steel, DIN, 1 m cable, HI 9835/HI 98360
663-5014
: Hanna
- Connection:DIN, 1 m cable
- For:HI 9835/HI 98360
- Type:HI 76309, 4-ring, built-in temperature sensor
- Description:Conductivity probe HI 76309, 4-ring, built-in temperature sensor, ATC
- Material:Stainless steel
- Seq:06