Neutral detergent, sterile, Klercide™ Neutral Detergent
Detergent for cleaning in critical areas to remove soiling prior to disinfection. Cleanroom (grade C) filtered to 0,2 µm, gamma irradiated and double bagged.
- Non-ionic with low residues
- Trigger spray pack is fitted with the SteriShield Delivery System™ to give 6 months in-use shelf life
- Available as a unit dose concentrate
It is designed to remove contamination in critical areas particularly cleanrooms, isolators, production equipment and all nonporous surfaces. It is particularly effective on oily and sticky spillages or following general maintenance especially where lubricants have been applied.
Frequently Bought Together