Order Entry
United Kingdom
ContactUsLinkComponent
Biocide, sterile, filtered, Klercide™ Sporicidal Chlorine/Quat
  148-0070
undefined
Biocide, sterile, filtered, Klercide™ Sporicidal Chlorine/Quat
  148-0070
  • Description:
    Klercide™ Sporicidal Chlorine/Quat, sterile filtered biocide, spray
  • Seq:
    01
  • Packed:
    1 L bottle with trigger spray
  • Pk:
    6
Klercide™ Sporicidal Chlorine/Quat is a fast acting sporicide. Suitable for ISO Class 5; Grade A and B (microbial count) areas: cleanroom processed and sterile, double bagged. Blend of stabilised chlorine dioxide and quaternary ammonium compound, with broad spectrum activity. Filtered to 0,2 µm.
  • Safe: non toxic and non corrosive, and very effective against bacterial spore
  • CEN and isolate tested
  • Fully compatible and ideal for a rotational regime
  • SteriShield Delivery System™ ensures 6 months in-use shelf life
 : EN standards: EN 1276 - bactericidal activity, EN 1650 - fungicidal activity, EN 13697 bactericidal surface test, EN 13704 sporocidal activity.
 : Use biocides safely. Always read the label and product information before use.