Order Entry
United Kingdom
ContactUsLinkComponent
2,5-Diethyl-1,3,4-thiadiazole for synthesis
2,5-Diethyl-1,3,4-thiadiazole for synthesis
Catalog # 8.15100.0025
Supplier:  Merck
CAS Number:  
2,5-Diethyl-1,3,4-thiadiazole for synthesis
Catalog # 8.15100.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 ml
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    (20 °C) slightly soluble
  • Molar Mass:
    142.22 g/mol
  • Boiling Point:
    105 °C (18.7 hPa)
  • Density:
    1.071 g/cm3 (20 °C)

Specification Test Results

About this item

 :
 :142.22 g/mol
 :M-Clarity™ quality level = MQ200
EINECS:255-143-3
 :GHS07
Storage Temperature:Ambient
CAS Number:40928-84-5
Formula:C₆H₁₀N₂S