Bismut(III) gallate basic hydrate ≥98% (by complexometric analysis, on dry substance), Sigma-Aldrich®
Catalog # SIAL15102-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: Bismuth subgallate hydrate, 2,7-Dihydroxy-1,3,2-benzodioxabismole-5-carboxylic acid hydrate, Dermatol hydrate
:
:394.09 g/mol (anhy)
EINECS:202-742-2
CAS Number:99-26-3
Formula:C₇H₅BiO₆·xH₂O
MDL Number:MFCD20441998