tert-Butyl hydroperoxide 5-6 mol/l in n-decane, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:75-91-2
:
Formula:C₄H₁₀O₂
MDL Number:MFCD00002130
:90.12 g/mol
ADR:5.2,
:M-Clarity™ quality level = MQ200
EINECS:200-915-7
UN:3109
:GHS02
:GHS05
:GHS06
:GHS08
:GHS09
Storage Temperature:Refrigerator
Frequently Bought Together