Curcumin
Catalog # EHERC11780000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,25 g
- Seq:3029
Specifications
About this item
:
:368.39 g/mol
EINECS:207-280-5
CAS Number:458-37-7
Formula:C₂₁H₂₀O₆
MDL Number:MFCD00008365