Order Entry
United Kingdom
ContactUsLinkComponent
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98% free radical
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98% free radical
Catalog # ABCRAB173085.0001
Supplier:  abcr
CAS Number:  
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98% free radical
Catalog # ABCRAB173085.0001
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :186.27 g/mol
Melting Pt:39…41 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:95407-69-5
Formula:C₁₀H₂₀NO₂
MDL Number:MFCD00270334