L(+)-Ornithine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3184-13-2
:
Formula:C₅H₁₂N₂O₂·HCl
MDL Number:MFCD00064562
:168.62 g/mol
:M-Clarity™ quality level = MQ200
EINECS:221-678-6
Frequently Bought Together