Cadmium(II) chloride hemipentahydrate, Sigma-Aldrich®
Catalog # SIAL208299-50G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:228.35 g/mol
Merck Index:13,01617
Melting Pt:568 °C
Density:2.84 g/cm³ (20 °C)
EINECS:233-296-7
UN:2570
ADR:6.1,III
:M-Clarity™ quality level = MQ100
:GHS06
:GHS08
:GHS09
CAS Number:7790-78-5
Formula:CdCl₂·2.5H₂O
MDL Number:MFCD00799071