Order Entry
United Kingdom
ContactUsLinkComponent
Overalls, Tyvek® 500 Labo, model CHF7
Overalls, Tyvek® 500 Labo, model CHF7
Supplier:  DuPont
Certificates
undefined
Overalls, Tyvek® 500 Labo, model CHF7
Supplier:  DuPont
Specifications
  • Catalog No:
  • 113-1551
  • 113-1552
  • 113-1547
  • 113-1548
  • 113-1549
  • 113-1550
  • Model:
  • 500 Labo CHF7
  • 500 Labo CHF7
  • 500 Labo CHF7
  • 500 Labo CHF7
  • 500 Labo CHF7
  • 500 Labo CHF7
  • Anti-static:
  • +
  • +
  • +
  • +
  • +
  • +
  • Cleanroom class:
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • 10000/ISO 7
  • Garment application:
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Cleanroom
  • Weight:
  • 41,5 g/m²
  • 41,5 g/m²
  • 41,5 g/m²
  • 41,5 g/m²
  • 41,5 g/m²
  • 41,5 g/m²