Benzyl carbazate
Marketsource
Supplier: BACHEM BIOCHEMICA
Jump to:
About this item
Synonyms: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate
- CAS Number:5331-43-1
- Formula:C₈H₁₀N₂O₂
- MDL Number:MFCD00041890
- Molecular Weight:166.18 g/mol
- EINECS:226-230-3
2 Options Available Below
Product Details & Documents
Synonyms: Hydrazine, N-CBZ protected, Benzyl hydrazinecarboxylate
- CAS Number:5331-43-1
- Formula:C₈H₁₀N₂O₂
- MDL Number:MFCD00041890
- Molecular Weight:166.18 g/mol
- EINECS:226-230-3
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



