

Acetyl chloride 1 M in dichloromethane
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-36-5
:
Formula:C₂H₃ClO
MDL Number:MFCD00000719
:78.5 g/mol
Density:1.294
Storage Temperature:Ambient
EINECS:200-865-6
UN:1593
ADR:6.1,III
:GHS08
:GHS07
Related Information
Frequently Bought Together