3,4-Diaminopyridine
Glass bottle for solids, 5 g
Catalog # MOLEM42199368
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3,4-Pyridinediamine, Amifampridine, 3,4-DAP, Pyridine-3,4-diyldiamine, Pyridine-3,4-diamine
- CAS Number:54-96-6
- Formula:C₅H₇N₃
- MDL Number:MFCD00006401
- Molecular Weight:109.13 g/mol
- Merck Index:13,03006
- Boiling Pt:262 °C (1013 hPa)
- Melting Pt:218…219 °C
- Flash Pt:240 °C
- Storage Temperature:Ambient
- EINECS:200-220-9
- UN:2811
- ADR:6.1,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



