Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,6-Dimethylnaphthalene
2,6-Dimethylnaphthalene
2,6-Dimethylnaphthalene
EHERC20775000
:
EHRENSTORFER
:
2,6-Dimethylnaphthalene
EHERC20775000
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
10 mg
Seq:
2275
:
:
156.23 g/mol
:
169.8 °C (1013 hPa)
:
–44.7 °C
:
1.8682 g/cm³ (20 °C)
:
Ambient
:
209-464-0
:
581-42-0
:
C₁₂H₁₂
:
MFCD00004120