Order Entry
United Kingdom
ContactUsLinkComponent
Zirconium(IV) oxide 98.5%
Irritant
Zirconium(IV) oxide 98.5%
Certificates
undefined
Irritant
Zirconium(IV) oxide 98.5%

Specification Test Results

  • Appearance:
    White Powder
  • Assay:
    ≥98.0 % (ZrO2 + HfO2)

Specification Test Results

Related Information

Frequently Bought Together