Cadmium selenide ≥99.995% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1306-24-7
:
Formula:CdSe
MDL Number:MFCD00010917
:191.37 g/mol
Merck Index:14,1626
Melting Pt:>1350 °C
Density:5.81
Storage Temperature:Ambient
EINECS:215-148-3
UN:3283
ADR:6.1,III
:GHS06
:GHS08
Frequently Bought Together