Order Entry
United Kingdom
ContactUsLinkComponent
Cadmium selenide ≥99.995% (metals basis)
Toxic
Health
Cadmium selenide ≥99.995% (metals basis)
Catalog # 11057.18
CAS Number:  
Toxic
Health
Cadmium selenide ≥99.995% (metals basis)
Catalog # 11057.18
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :191.37 g/mol
Merck Index:14,1626
Melting Pt:>1350 °C
Density:5.81
Storage Temperature:Ambient
EINECS:215-148-3
UN:3283
ADR:6.1,III
 :GHS06
 :GHS08
CAS Number:1306-24-7
Formula:CdSe
MDL Number:MFCD00010917