Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
 

Silver(I) oxide ≥99% (metals basis)

Oxidizing
Corrosive
Silver(I) oxide ≥99% (metals basis)
Certificates
Silver(I) oxide ≥99% (metals basis)
Silver(I) oxide ≥99% (metals basis)

3 Options Available Below

Product Details & Documents

Purity: 99+% CAS No.: 20667-12-3 Molecular Formula: Ag2O Formula Weight: 231.74 Physical Form: Powder Density: 7.2 Appearance: Dark brown Odour: Odourless Melting Point: 230° dec. Merck Reference: 14,8521 UN No.: 1479 MDL No.: MFCD00003404

Polishing glass, Colouring glass yellow, catalyst, purifying drinking water, lab reagent

Synonyms:  di-Silver oxide, Disilver oxide, Silver oxide
  • CAS Number:20667-12-3
  • Formula:Ag₂O
  • MDL Number:MFCD00003404
  • Molecular Weight:231.74 g/mol
  • Merck Index:14,8521
  • Melting Pt:230 °C (dec.)
  • Density:7.2
  • Storage Temperature:Ambient
  • EINECS:243-957-1
  • UN:1479
  • ADR:5.1,II
Product Family Options

Product Information