Lanthanum(III) oxide 99.9% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1312-81-8
:
Formula:La₂O₃
MDL Number:MFCD00011071
:325.81 g/mol
Boiling Pt:4200 °C (760 mmHg)
Melting Pt:2315 °C
Density:6,51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-200-5
Specification Test Results
- Appearance:White to off-white Powder
- Assay:≥99.85 % (La2O3) TREO base
- Loss on ignition:≤3.0 % (1000°C)
- Iron(III)oxide (Fe2O3):≤250 ppm
Specification Test Results
Frequently Bought Together