Palladium 10% on activated carbon wetted with ca. 50% water hydrogenation catalyst, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7440-05-3
:
Formula:Pd
MDL Number:MFCD00011167
:106.420 g/mol
ADR:4.1,III
:Moistened with water
EINECS:231-115-6
Frequently Bought Together