Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Triethanolamine hydrochloride

Triethanolamine hydrochloride

Plastic bottle for solids, 250 g
Catalog # SERA36885.02
Marketsource
Supplier:   Serva
Triethanolamine hydrochloride
Catalog # SERA36885.02
Supplier:  Serva
Marketsource
CAS Number:  637-39-8
Triethanolamine hydrochloride

Plastic bottle for solids, 250 g

CAS Number:  637-39-8
Price
Inquire For Price
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:637-39-8
  • Formula:C₆H₁₆ClNO₃
  • MDL Number:MFCD00012596
  • Molecular Weight:185.65 g/mol
  • Storage Temperature:Ambient
  • EINECS:211-284-2
Specifications

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.