Triethanolamine hydrochloride
Glass bottle for solids, 100 g
Catalog # ROTH4804.1
Marketsource
Supplier: Roth Carl
Triethanolamine hydrochloride
Catalog # ROTH4804.1
Supplier: Roth Carl
Marketsource
CAS Number: 637-39-8
Jump to:
Product Details & Documents
- CAS Number:637-39-8
- Formula:C₆H₁₆ClNO₃
- MDL Number:MFCD00012596
- Molecular Weight:185.65 g/mol
- Storage Temperature:Ambient
- EINECS:211-284-2
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



