Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
4'-Ethoxyacetanilide

4'-Ethoxyacetanilide

Glass bottle for solids, 1 g
Catalog # PROOICRS0380
4'-Ethoxyacetanilide
Catalog # PROOICRS0380
CAS Number:  62-44-2
4'-Ethoxyacetanilide

Glass bottle for solids, 1 g

CAS Number:  62-44-2
Price
£337.00
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Acetphenetidin, Acetophenetidin, Phenacetin, N-(4-Ethoxyphenyl)acetamide, 4-Acetophenetidide
  • CAS Number:62-44-2
  • Formula:C₁₀H₁₃NO₂
  • MDL Number:MFCD00009094
  • Molecular Weight:179.22 g/mol
  • EINECS:200-533-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.