


Diallylamine
Supplier: Merck
Certificates
About this item
CAS Number:124-02-7
:
Formula:C₆H₁₁N
MDL Number:MFCD00008642
ADR:3 (6.1, 8),II
:M-Clarity™ quality level = MQ200
:97.16 g/mol
EINECS:204-671-2
UN:2359
:GHS02
:GHS06
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Solubility:86 g/l (20 °C)
- Melting Point:-88 °C
- Molar Mass:97.15 g/mol
- Boiling Point:111 °C (1013 hPa)
- Vapor Pressure:24.2 hPa (20 °C)
- Flash Point:11 °C
- Refractive Index:1.4405 (20 °C, 589 nm)
- Explosion Limit:0.93 - 14.2 %(V)
- Density:0.79 g/cm3 (20 °C)
- pH:11.5 (9.7 g/l, H2O)
- Ignition Point:273 °C
Specification Test Results
Frequently Bought Together