Order Entry
United Kingdom
ContactUsLinkComponent
3-Chloropropene stabilised, Sigma-Aldrich®
Flammable
Health
Irritant
Environment
3-Chloropropene stabilised, Sigma-Aldrich®
Supplier:  Merck
Certificates
Flammable
Health
Irritant
Environment
3-Chloropropene stabilised, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    3.6 g/l (20 °C)
  • Melting Point:
    -136 °C
  • Molar Mass:
    76.52 g/mol
  • Boiling Point:
    45 °C (1013 hPa)
  • Vapor Pressure:
    400 hPa (20 °C)
  • Explosion Limit:
    3.3 - 11.2 %(V)
  • Density:
    0.94 g/cm3 (20 °C)
  • pH:
    (H2O) not applicable
  • Ignition Point:
    390 °C

Specification Test Results

Related Information

Frequently Bought Together