Pesticide reference standard, Nitroguanidine 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA15588000AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:556-88-7
- Description:Nitroguanidine 100 µg/ml in acetonitrile
- Formula:CH₄N₄O₂
- Description:Nitroguanidine