Triethanolamine hydrochloride ≥99%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99+% CAS No.: 637-39-8 Molecular Formula: C6H15NO3.HCl Formula Weight: 185.65 Melting Point: 177-179° Beilstein Reference: 3909940 Merck Reference: 14,9665 MDL No.: MFCD00012596
- CAS Number:637-39-8
- Formula:C₆H₁₆ClNO₃
- MDL Number:MFCD00012596
- Molecular Weight:185.65 g/mol
- Merck Index:14,9665
- Melting Pt:177…179 °C
- Storage Temperature:Ambient
- EINECS:211-284-2
3 Options Available Below
Product Details & Documents
Purity: 99+% CAS No.: 637-39-8 Molecular Formula: C6H15NO3.HCl Formula Weight: 185.65 Melting Point: 177-179° Beilstein Reference: 3909940 Merck Reference: 14,9665 MDL No.: MFCD00012596
- CAS Number:637-39-8
- Formula:C₆H₁₆ClNO₃
- MDL Number:MFCD00012596
- Molecular Weight:185.65 g/mol
- Merck Index:14,9665
- Melting Pt:177…179 °C
- Storage Temperature:Ambient
- EINECS:211-284-2



