3,4-Dihydro-2H-pyran 99%
:110-87-2
:
:C₅H₈O
:MFCD00006558
:0.92 g/cm³
:84.12 g/mol
:–70 °C
:–9 °C
:Ambient
:203-810-4
:2376
:3 II
:GHS07
:GHS02
- Environmentally Preferable:
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4395 to 1.4435 (20°C, 589 nm)
- GC:≥98.5 %
Frequently Bought Together