(S)-(+)-β-Aminophenethyl alcohol
Catalog # ABCRAB168529.0025
Supplier: abcr
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
About this item
:
:137.18 g/mol
CAS Number:20989-17-7
Formula:C₈H₁₁NO
MDL Number:MFCD00064404