
Calcium chloride, dried 97%, powder
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10043-52-4
:
Formula:CaCl₂
MDL Number:MFCD00010903
Merck Index:14,1659
Boiling Pt:>1600 °C
Melting Pt:782 °C
Density:2.174 g/cm³ (20 °C)
:110.98 g/mol
Storage Temperature:Ambient
EINECS:233-140-8
REACH:01-2119494219-28
:GHS07
Frequently Bought Together