Order Entry
United Kingdom
ContactUsLinkComponent
Sodium orthovanadate, Millipore®
Irritant
Sodium orthovanadate, Millipore®
Catalog # 567540-5
CAS Number:  
Irritant
Sodium orthovanadate, Millipore®
Catalog # 567540-5
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :183.91 g/mol
Melting Pt:850…866 °C
EINECS:237-287-9
 :GHS07
Storage Temperature:Ambient
CAS Number:13721-39-6
Formula:Na₃O₄V
MDL Number:MFCD00003511