Sodium hexafluorosilicate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16893-85-9
:
Formula:F₆Na₂Si
MDL Number:MFCD00003491
:188.06 g/mol
Merck Index:14,8624
Melting Pt: Dec.
Density:2.679
Storage Temperature:Ambient
EINECS:240-934-8
UN:2674
ADR:6.1,III
:GHS06
Related Information
Frequently Bought Together