Sodium selenite, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10102-18-8
:
Formula:Na₂SeO₃
MDL Number:MFCD00003489
Merck Index:12,08822
Melting Pt:710 °C
Density:3.1 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:6.1,I
:M-Clarity™ quality level = MQ200
:172.94 g/mol
EINECS:233-267-9
UN:2630
:GHS06
:GHS09
Frequently Bought Together