

Sodium hexafluoroantimonate ≥99.5%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16925-25-0
:
Formula:F₆NaSb
MDL Number:MFCD00003482
:258.74 g/mol
Density:3.38
Storage Temperature:Ambient
EINECS:240-989-8
UN:1549
ADR:6.1,III
:GHS07
:GHS09
Related Information
Frequently Bought Together