Order Entry
United Kingdom
ContactUsLinkComponent
Sodium hexafluoroantimonate 98%
Irritant
Environment
Sodium hexafluoroantimonate 98%
Certificates
undefined
Irritant
Environment
Sodium hexafluoroantimonate 98%

Specification Test Results

  • Appearance:
    White to off-white Crystalline powder
  • Assay:
    ≥97.5 %
  • Melting point:
    80 to 84 °C
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together