


Sodium peroxodisulphate
Catalog # ROTH4365.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
Specifications
About this item
:
:238.11 g/mol
Merck Index:13,08729
Melting Pt:100 °C
Density:2.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-892-1
UN:1505
ADR:5.1,III
:GHS03
:GHS08
:GHS07
CAS Number:7775-27-1
Formula:Na₂S₂O₈
MDL Number:MFCD00003501