Water USP, JP, Ph. Eur., purified
: Hydrogen oxide
:7732-18-5
:
:H₂O
:MFCD00011332
:100
:0
:1.00 g/cm³ (20 °C)
:18.02 g/mol
:13,10098
:Ambient
:231-791-2
- Environmentally Preferable:
- Appearance:Clear colourless liquid
- Heavy metals (as Pb):Max. 0.1 ppm
- Total organic carbon (TOC) (on line):Max. 50 ppb
- Yeasts and moulds (total count):Max. 100 CFU/ml
- Bacterial endotoxins (IU/ml):Max. 0.25
- Residual solvents (CPMP/ICH/283/95):Unlikely by manuf.process
- Residues of metal catalysts or reagents:Unlikely by manuf.process
- As (Arsenic):Max. 0.1 ppm
- Cd (Cadmium):Max. 0.1 ppm
- Co (Cobalt):Max. 0.1 ppm
- Hg (Mercury):Max. 0.1 ppm
- Ni (Nickel):Max. 0.1 ppm
- Pb (Lead):Max. 0.1 ppm
- V (Vanadium):Max. 0.1 ppm
- Conforms to Ph.Eur./USP/JP:Passes test
- Conforms to Ph.Eur. Monograph 0008:Passes test
- Total organic carbon (TOC) [Ph.Eur.]:Max. 0.5 mg/l
- Conductivity (20°C) (off line) [Ph.Eur.]:≤ 1.1 µS/cm
- Elemental impurities (by conductivity):Passes test
- Conductivity (20°C) (on line) [Ph.Eur.]:Max. 1.1 μS/cm
- Acidity or alkalinity:Passes test Ph.Eur.
- Oxidisable substances:Passes test Ph.Eur.
- Cl (Chloride):Passes test Ph.Eur.
- SO4 (Sulphate):Passes test Ph.Eur.
- NH4 (Ammonium) [Ph.Eur.]:Max. 0.2 ppm
- Calcium and magnesium:Passes test Ph.Eur.
- Residue on evaporation [Ph.Eur.]:Max. 0.001 %
- Microbial contamination:Passes test Ph.Eur.
- TAMC [Ph.Eur.]:Max. 100 CFU/ml
- Total organic carbon [USP]:Max. 0.50 mg/l
- Water conductivity [USP]:Max. 1.1 μS/cm
- Total organic carbon [JP]:Max. 0.50 mg/l
- Conductivity (25°C) [JP]:Max. 2.1 μS/cm
- KMnO4 reducing substances:Passes test JP
- Microbial limit:Passes test JP
- TAMC [JP]:Max. 100 CFU/ml
- Bacterial endotoxins (Max. 0,25 IU/ml), tested on finished product:Passes test
1000L HDPE IBC W UN pallet (ZPACK13960015) 10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 200L Plastic drums for liquids UN (ZPACK13950512) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
90200_WATER PURIFIED PH.EUR USP NF_DIOXIN_REV1 90200_WATER PURIFIED PH.EUR USP NF_MELAMINE.REV1 VWRC 90200 Water purified PH.EUR.USPNFJP Water Ph. Eur., USP, JP, Purified
90200_WATER PURIFIED PH.EUR USP NF_MANUFFLOW.pdf Maufacturing Processflow_90200_Water purified PH.EUR.USPNFJP
Frequently Bought Together