Sodium nitrite ≥99.0%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur.
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7632-00-0
:
Formula:NaNO₂
MDL Number:MFCD00011118
:69 g/mol
Merck Index:13,08721
Boiling Pt:320 °C (1013 hPa)
Melting Pt:284 °C
Density:2.168 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-555-9
UN:1500
ADR:5.1 (6.1),III
:GHS03
:GHS06
:GHS09
Specification Test Results
- Assay:Min. 99.0 %
- Heavy metals (as Pb):Max. 10 ppm
- Insolubility in water:Max. 100 ppm
- Cl (Chloride):Max. 50 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ca (Calcium):Max. 20 ppm
- Fe (Iron):Max. 10 ppm
- K (Potassium):Max. 50 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 250 ml Plastic bottle for solids DIN 60 (ZPACK13250250)
Packaging Information
27960_SODIUM NITRITE ANALAR NORMAPUR_BSE_REV0
BSE/TSE Certificates
spec_VWRC27960.236 spec_VWRC27960.236 spec_VWRC27960.293
Product specification
Frequently Bought Together