Sodium iodide ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent
:7681-82-5
:
:NaI
:MFCD00003532
:149.89 g/mol
:13,08705
:1300 °C (1013 hPa)
:662 °C
:3.67 g/cm³ (20 °C)
:Ambient
:231-679-3
:3077
:9,III
:GHS09
- Assay:Min. 99.5 %
- pH (25°C; 5 %):6.0 - 9.0
- Heavy metals (as Pb):Max. 5 ppm
- Insoluble matter:Max. 100 ppm
- Loss on drying (100-105°C):Max. 0.5 %
- Total N (Nitrogen):Max. 40 ppm
- Cl + Br (as Cl):Max. 100 ppm
- IO3 (Iodate):Max. 3 ppm
- PO4 (Phosphate):Max. 10 ppm
- SO4 (Sulphate):Max. 50 ppm
- Ba (Barium):Max. 20 ppm
- Ca (Calcium):Max. 20 ppm
- Cu (Copper):Max. 5 ppm
- Fe (Iron):Max. 5 ppm
- K (Potassium):Max. 100 ppm
- Mg (Magnesium):Max. 10 ppm
- Ni (Nickel):Max. 1 ppm
- Conforms to ACS:Passes test
Frequently Bought Together