Order Entry
United Kingdom
ContactUsLinkComponent
Sodium iodide, anhydrous ≥99% (H2O < 1%)
Sodium iodide, anhydrous ≥99% (H2O < 1%)
Catalog # ABCRAB116701.0100
Supplier:  abcr
CAS Number:  
Sodium iodide, anhydrous ≥99% (H2O < 1%)
Catalog # ABCRAB116701.0100
Supplier:  abcr
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

About this item

 :
 :149.89 g/mol
Merck Index:13,08705
Boiling Pt:1304 °C
Melting Pt:651 °C
Density:3.667
Storage Temperature:Ambient
EINECS:231-679-3
UN:3077
ADR:9,III
 :GHS07
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532