Bottles, wide neck, with screw cap, BOLA
PTFE, white.
- Temperature resistance: from −200 to +250 °C
- Universal chemical resistance
- Pressure: no pressure
Thick-walled, smooth interior surface, with screw cap.
Frequently Bought Together
PTFE, white.
Thick-walled, smooth interior surface, with screw cap.